5H-Dibenzo[a,d][7]annulen-5-one oxime 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD262795-250MGEA 489 DKK
BLDPBD262795-250MG BLDPBD262795-1G BLDPBD262795-100MG BLDPBD262795-5G
5H-Dibenzo[a,d][7]annulen-5-one oxime 95+%
5H-Dibenzo[a,d][7]annulen-5-one oxime

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₁₁NO
Molekylvægt: 221,25 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00021100
CAS-nummer: 1021-91-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR