3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-amine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD262775-5GEA 3260 DKK
BLDPBD262775-5G BLDPBD262775-1G BLDPBD262775-250MG BLDPBD262775-100MG
3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-amine 95%
3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydronaphthalen-2-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₅H₂₃N
Molekylvægt: 217,35 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22572337
CAS-nummer: 116233-17-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR