5-Chloro-3-(trifluoromethyl)-1H-pyrazole 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD26262-250MGEA 2760 DKK
BLDPBD26262-250MG BLDPBD26262-1G BLDPBD26262-100MG
5-Chloro-3-(trifluoromethyl)-1H-pyrazole 95%
5-Chloro-3-(trifluoromethyl)-1H-pyrazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₄H₂ClF₃N₂
Molekylvægt: 170,52 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07375186
CAS-nummer: 131797-35-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR