2-(4-Methoxyphenyl)cyclopropanecarboxylic acid 98+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD262532-100MGEA 458 DKK
BLDPBD262532-100MG BLDPBD262532-250MG BLDPBD262532-1G
2-(4-Methoxyphenyl)cyclopropanecarboxylic acid 98+%
2-(4-Methoxyphenyl)cyclopropanecarboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₁₂O₃
Molekylvægt: 192,21 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09834141
CAS-nummer: 92016-94-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR