5-Chloro-2-fluorobenzimidamide hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD26244-5GEA 5650 DKK
BLDPBD26244-5G BLDPBD26244-250MG BLDPBD26244-1G
5-Chloro-2-fluorobenzimidamide hydrochloride 97%
5-Chloro-2-fluorobenzimidamide hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₇Cl₂FN₂
Molekylvægt: 209,04 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04973393
CAS-nummer: 1187929-52-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR