Decaethylene glycol monomethyl ether 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD262364-250MGEA 303 DKK
BLDPBD262364-250MG BLDPBD262364-1G BLDPBD262364-100MG
Decaethylene glycol monomethyl ether 95%
Decaethylene glycol monomethyl ether

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₁H₄₄O₁₁
Molekylvægt: 472,57 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06797087
CAS-nummer: 27425-92-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR