3,6,9,12,15,18,21,24,27,30-Decaoxadotriacontane-1,32-diol 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD262354-250MGEA 210 DKK
BLDPBD262354-250MG BLDPBD262354-5G BLDPBD262354-1G BLDPBD262354-100MG
3,6,9,12,15,18,21,24,27,30-Decaoxadotriacontane-1,32-diol 98%
3,6,9,12,15,18,21,24,27,30-Decaoxadotriacontane-1,32-diol

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₂H₄₆O₁₂
Molekylvægt: 502,59 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD26793839
CAS-nummer: 6809-70-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR