(S)-2-Phenylpyrrolidine hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD262265-100MGEA 636 DKK
BLDPBD262265-100MG
(S)-2-Phenylpyrrolidine hydrochloride 95%
(S)-2-Phenylpyrrolidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₄ClN
Molekylvægt: 183,67 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12910597
CAS-nummer: 56523-58-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR