Methyl-5-bromo-4-methylpicolinate 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD26221-10GEA 5350 DKK
BLDPBD26221-10G BLDPBD26221-250MG BLDPBD26221-1G BLDPBD26221-5G BLDPBD26221-25G BLDPBD26221-100MG
Methyl-5-bromo-4-methylpicolinate 96%
Methyl-5-bromo-4-methylpicolinate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₈BrNO₂
Molekylvægt: 230,0600 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07375102
CAS-nummer: 886365-06-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR