4,7,10,13-Tetraoxahexadecane-1,16-dinitrile 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD262155-1GEA 1350 DKK
BLDPBD262155-1G BLDPBD262155-250MG BLDPBD262155-100MG
4,7,10,13-Tetraoxahexadecane-1,16-dinitrile 98%
4,7,10,13-Tetraoxahexadecane-1,16-dinitrile

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₂₀N₂O₄
Molekylvægt: 256,29 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00483880
CAS-nummer: 57741-46-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR