5-Bromo-2-ethoxypyrimidine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD26190-10GEA 907 DKK
BLDPBD26190-10G BLDPBD26190-5G BLDPBD26190-25G BLDPBD26190-1G
5-Bromo-2-ethoxypyrimidine 98%
5-Bromo-2-ethoxypyrimidine

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₇BrN₂O
Molekylvægt: 203,0400 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07375135
CAS-nummer: 17758-11-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR