1-Benzyl-3,3-difluoropyrrolidine hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD261879-100MGEA 667 DKK
BLDPBD261879-100MG BLDPBD261879-250MG
1-Benzyl-3,3-difluoropyrrolidine hydrochloride 95%
1-Benzyl-3,3-difluoropyrrolidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₄ClF₂N
Molekylvægt: 233,69 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14706680
CAS-nummer: 862416-37-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR