4-(Trifluoromethyl)pyridine-2,6-diol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD261698-1GEA 1410 DKK
BLDPBD261698-1G BLDPBD261698-100MG BLDPBD261698-250MG
4-(Trifluoromethyl)pyridine-2,6-diol 95%
4-(Trifluoromethyl)pyridine-2,6-diol

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₄F₃NO₂
Molekylvægt: 179,09 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11100755
CAS-nummer: 356518-28-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR