5-Bromo-1,3-dimethyl-1H-pyrazole 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD26164-100MGEA 365 DKK
BLDPBD26164-100MG BLDPBD26164-1G BLDPBD26164-250MG BLDPBD26164-5G
5-Bromo-1,3-dimethyl-1H-pyrazole 97%
5-Bromo-1,3-dimethyl-1H-pyrazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₅H₇BrN₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08448181
CAS-nummer: 5744-70-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR