Pyridin-2-ylmethyl 2-(4-isobutylphenyl)propanoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD261573-5GEA 3210 DKK
BLDPBD261573-5G BLDPBD261573-25G BLDPBD261573-1G BLDPBD261573-0.025 BLDPBD261573-250MG BLDPBD261573-10G
Pyridin-2-ylmethyl 2-(4-isobutylphenyl)propanoate 95%
Pyridin-2-ylmethyl 2-(4-isobutylphenyl)propanoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₉H₂₃NO₂
Molekylvægt: 297,39 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00866018
CAS-nummer: 64622-45-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR