5-Methyl-1H-indazol-6-ol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD261418-5GEA 11190 DKK
BLDPBD261418-5G BLDPBD261418-100MG BLDPBD261418-1G BLDPBD261418-250MG
5-Methyl-1H-indazol-6-ol 95%
5-Methyl-1H-indazol-6-ol

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈N₂O
Molekylvægt: 148,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11007971
CAS-nummer: 1082042-15-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR