1-(Dimethylamino)-6-methylheptan-3-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD261156-5GEA 7450 DKK
BLDPBD261156-5G BLDPBD261156-1G BLDPBD261156-100MG BLDPBD261156-250MG
1-(Dimethylamino)-6-methylheptan-3-one 95%
1-(Dimethylamino)-6-methylheptan-3-one

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₁NO
Molekylvægt: 171,28 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22683883
CAS-nummer: 107245-26-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR