4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))tribenzoic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD261054-1GEA 2480 DKK
BLDPBD261054-1G BLDPBD261054-100MG BLDPBD261054-5G BLDPBD261054-250MG
4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))tribenzoic acid 98%
4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))tribenzoic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃₃H₁₈O₆
Molekylvægt: 510,49 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD17926334
CAS-nummer: 205383-17-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR