tert-Butyl 6-hydroxy-2,3-dihydro-1H-indole-1-carboxylate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD260963-1GEA 512 DKK
BLDPBD260963-1G BLDPBD260963-250MG BLDPBD260963-100MG
tert-Butyl 6-hydroxy-2,3-dihydro-1H-indole-1-carboxylate 97%
tert-Butyl 6-hydroxy-2,3-dihydro-1H-indole-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₃H₁₇NO₃
Molekylvægt: 235,279 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11045243
CAS-nummer: 957204-30-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR