4-[(N,N-Dimethylamino)methyl]benzeneboronic acid pinacol ester hydrochloride 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD260905-1GEA 698 DKK
BLDPBD260905-1G BLDPBD260905-5G BLDPBD260905-250MG
4-[(N,N-Dimethylamino)methyl]benzeneboronic acid pinacol ester hydrochloride 98%
4-[(N,N-Dimethylamino)methyl]benzeneboronic acid pinacol ester hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₅H₂₄BNO₂·ClH
Molekylvægt: 297,628 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07771993
CAS-nummer: 1073371-85-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR