2-Amino-5-iodo-3-nitropyridine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD2609-5GEA 334 DKK
BLDPBD2609-5G BLDPBD2609-1G BLDPBD2609-25G
2-Amino-5-iodo-3-nitropyridine 97%
2-Amino-5-iodo-3-nitropyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₅H₄IN₃O₂
Molekylvægt: 265,009 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04038464
CAS-nummer: 25391-57-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR