trans-4-Morpholinocyclohexanol hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD260815-250MGEA 915 DKK
BLDPBD260815-250MG BLDPBD260815-100MG BLDPBD260815-1G
trans-4-Morpholinocyclohexanol hydrochloride 97%
trans-4-Morpholinocyclohexanol hydrochloride

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₀ClNO₂
Molekylvægt: 221,72 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26940271
CAS-nummer: 1588441-09-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR