Diethyl 3-fluorophthalate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD260655-1GEA 915 DKK
BLDPBD260655-1G BLDPBD260655-5G BLDPBD260655-250MG
Diethyl 3-fluorophthalate 95%
Diethyl 3-fluorophthalate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₃FO₄
Molekylvægt: 240,23 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16037329
CAS-nummer: 65610-10-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR