Methyl 4-bromo-2-fluoro-6-methoxybenzoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD260525-250MGEA 814 DKK
BLDPBD260525-250MG BLDPBD260525-1G BLDPBD260525-100MG BLDPBD260525-5G
Methyl 4-bromo-2-fluoro-6-methoxybenzoate 95%
Methyl 4-bromo-2-fluoro-6-methoxybenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₈BrFO₃
Molekylvægt: 263,06 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD23707517
CAS-nummer: 1427415-25-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR