2-Amino-4-(3-pyridyl)thiazole 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD26043-1GEA 652 DKK
BLDPBD26043-1G BLDPBD26043-100MG BLDPBD26043-5G BLDPBD26043-250MG
2-Amino-4-(3-pyridyl)thiazole 97%
2-Amino-4-(3-pyridyl)thiazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₇N₃S
Molekylvægt: 177,22 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00218013
CAS-nummer: 30235-27-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR