1-Bromo-4-fluoroisoquinoline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD260372-100MGEA 1340 DKK
BLDPBD260372-100MG BLDPBD260372-1G BLDPBD260372-250MG
1-Bromo-4-fluoroisoquinoline 95%
1-Bromo-4-fluoroisoquinoline

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₅BrFN
Molekylvægt: 226,05 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD26406841
CAS-nummer: 1421517-86-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR