4-Chloro-1H-imidazole-2-carboxylic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD260228-100MGEA 853 DKK
BLDPBD260228-100MG BLDPBD260228-250MG
4-Chloro-1H-imidazole-2-carboxylic acid 97%
4-Chloro-1H-imidazole-2-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₄H₃ClN₂O₂
Molekylvægt: 146,53 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 1824084-44-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR