4-Bromo-8-(trifluoromethoxy)quinoline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259608-1GEA 1600 DKK
BLDPBD259608-1G BLDPBD259608-5G BLDPBD259608-250MG BLDPBD259608-100MG
4-Bromo-8-(trifluoromethoxy)quinoline 95%
4-Bromo-8-(trifluoromethoxy)quinoline

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₅BrF₃NO
Molekylvægt: 292,05 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12674964
CAS-nummer: 1189105-60-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR