4-Bromo-6-(trifluoromethoxy)quinoline 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259606-100MGEA 225 DKK
BLDPBD259606-100MG BLDPBD259606-250MG
4-Bromo-6-(trifluoromethoxy)quinoline 98%
4-Bromo-6-(trifluoromethoxy)quinoline

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₅BrF₃NO
Molekylvægt: 292,05 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12674963
CAS-nummer: 1189105-68-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR