2-Chlorothieno[3,2-c]pyridine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259599-250MGEA 2180 DKK
BLDPBD259599-250MG BLDPBD259599-100MG BLDPBD259599-1G
2-Chlorothieno[3,2-c]pyridine 95%
2-Chlorothieno[3,2-c]pyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₄ClNS
Molekylvægt: 169,63 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01320520
CAS-nummer: 28783-23-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR