tert-Butyl 4-ethynylbenzylcarbamate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259581-5GEA 1890 DKK
BLDPBD259581-5G BLDPBD259581-100MG BLDPBD259581-250MG BLDPBD259581-1G
tert-Butyl 4-ethynylbenzylcarbamate 95%
tert-Butyl 4-ethynylbenzylcarbamate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₄H₁₇NO₂
Molekylvægt: 231,29 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18207366
CAS-nummer: 680190-96-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR