Methyl 4-formyl-3-hydroxybenzoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259532-5GEA 4350 DKK
BLDPBD259532-5G BLDPBD259532-1G BLDPBD259532-100MG BLDPBD259532-50MG BLDPBD259532-250MG
Methyl 4-formyl-3-hydroxybenzoate 95%
Methyl 4-formyl-3-hydroxybenzoate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₈O₄
Molekylvægt: 180,15 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD17015455
CAS-nummer: 24589-98-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR