3-(Bromomethyl)-4-methylpyridine hydrobromide 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259531-100MGEA 861 DKK
BLDPBD259531-100MG BLDPBD259531-1G BLDPBD259531-250MG
3-(Bromomethyl)-4-methylpyridine hydrobromide 97%
3-(Bromomethyl)-4-methylpyridine hydrobromide

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₉Br₂N
Molekylvægt: 266,96 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD26743700
CAS-nummer: 1384972-83-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR