tert-Butyl (3-bromo-4-methylphenyl)carbamate 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259337-5GEA 597 DKK
BLDPBD259337-5G BLDPBD259337-1G BLDPBD259337-25G
tert-Butyl (3-bromo-4-methylphenyl)carbamate 96%
tert-Butyl (3-bromo-4-methylphenyl)carbamate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₆BrNO₂
Molekylvægt: 286,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12913704
CAS-nummer: 515813-02-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR