6-Chloro-n,n-dimethyl-3-pyridinamine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259319-250MGEA 512 DKK
BLDPBD259319-250MG
6-Chloro-n,n-dimethyl-3-pyridinamine 98%
6-Chloro-n,n-dimethyl-3-pyridinamine
Formel: C₇H₉ClN₂
Molekylvægt: 156,613 g/mol
MDL Number: MFCD00234337
CAS-nummer: 41288-91-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR