7-Bromo-4H-pyrido[1,2-a]pyrimidin-4-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259273-250MGEA 675 DKK
BLDPBD259273-250MG BLDPBD259273-100MG
7-Bromo-4H-pyrido[1,2-a]pyrimidin-4-one 95%
7-Bromo-4H-pyrido[1,2-a]pyrimidin-4-one

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₅BrN₂O
Molekylvægt: 225,04 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07434965
CAS-nummer: 23536-76-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR