(2-Amino-3-methoxyphenyl)methanol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259260-1GEA 1280 DKK
BLDPBD259260-1G BLDPBD259260-100MG BLDPBD259260-250MG
(2-Amino-3-methoxyphenyl)methanol 95%
(2-Amino-3-methoxyphenyl)methanol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₁₁NO₂
Molekylvægt: 153,17 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09029933
CAS-nummer: 205877-13-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR