6-Bromo-3-iodoquinolin-4-ol 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD259053-100MGEA 287 DKK
BLDPBD259053-100MG BLDPBD259053-250MG BLDPBD259053-1G
6-Bromo-3-iodoquinolin-4-ol 95+%
6-Bromo-3-iodoquinolin-4-ol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₅BrINO
Molekylvægt: 349,95 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD15527294
CAS-nummer: 1260886-58-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR