6-Chloro-4-formyl-nicotinic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD258980-250MGEA 1790 DKK
BLDPBD258980-250MG BLDPBD258980-1G BLDPBD258980-100MG
6-Chloro-4-formyl-nicotinic acid 95%
6-Chloro-4-formyl-nicotinic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₄ClNO₃
Molekylvægt: 185,565 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18258739
CAS-nummer: 1031433-06-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR