3,5-Dimethyl-1H-pyrazole-4-carbonitrile 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD258875-250MGEA 1850 DKK
BLDPBD258875-250MG BLDPBD258875-100MG
3,5-Dimethyl-1H-pyrazole-4-carbonitrile 95%
3,5-Dimethyl-1H-pyrazole-4-carbonitrile

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₇N₃
Molekylvægt: 121,13 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09260786
CAS-nummer: 108161-12-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR