4'-(Methylsulphonyl)-[1,1'-biphenyl]-2-carboxylic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD258834-250MGEA 148 DKK
BLDPBD258834-250MG
4'-(Methylsulphonyl)-[1,1'-biphenyl]-2-carboxylic acid 95%
4'-(Methylsulphonyl)-[1,1'-biphenyl]-2-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₂O₄S
Molekylvægt: 276,30 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06802579
CAS-nummer: 893736-90-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR