4-Bromo-3-fluoroaniline, N-CBZ protected 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD258647-1GEA 249 DKK
BLDPBD258647-1G BLDPBD258647-5G BLDPBD258647-250MG
4-Bromo-3-fluoroaniline, N-CBZ protected 97%
4-Bromo-3-fluoroaniline, N-CBZ protected

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₁BrFNO₂
Molekylvægt: 324,145 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD23381075
CAS-nummer: 510729-01-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR