2,3,4,5-Tetramethylaniline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD258588-1GEA 5000 DKK
BLDPBD258588-1G BLDPBD258588-50MG BLDPBD258588-100MG BLDPBD258588-250MG
2,3,4,5-Tetramethylaniline 95%
2,3,4,5-Tetramethylaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₀H₁₅N
Molekylvægt: 149,23 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD19203597
CAS-nummer: 2217-45-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR