4-Bromo-2-iodo-1-(phenylsulphonyl)-1H-pyrrolo[2,3-b]pyridine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD25855-100MGEA 931 DKK
BLDPBD25855-100MG BLDPBD25855-250MG
4-Bromo-2-iodo-1-(phenylsulphonyl)-1H-pyrrolo[2,3-b]pyridine 95%
4-Bromo-2-iodo-1-(phenylsulphonyl)-1H-pyrrolo[2,3-b]pyridine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₃H₈BrIN₂O₂S
Molekylvægt: 463,09 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08443802
CAS-nummer: 889939-26-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR