4,5-Diamino-2-fluorobenzotrifluoride 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD258123-10GEA 7380 DKK
BLDPBD258123-10G BLDPBD258123-100MG BLDPBD258123-250MG BLDPBD258123-5G BLDPBD258123-1G
4,5-Diamino-2-fluorobenzotrifluoride 98%
4,5-Diamino-2-fluorobenzotrifluoride

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₆F₄N₂
Molekylvægt: 194,13 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04038250
CAS-nummer: 179062-06-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR