5-Fluoro-3-methylsalicylaldehyde 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD258122-5GEA 962 DKK
BLDPBD258122-5G BLDPBD258122-1G BLDPBD258122-250MG BLDPBD258122-25G
5-Fluoro-3-methylsalicylaldehyde 95%
5-Fluoro-3-methylsalicylaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₇FO₂
Molekylvægt: 154,1384 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04039244
CAS-nummer: 704884-74-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR