4,5,6,7-Tetrahydro-1H-pyrazolo[3,4-c]pyridine dihydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD25810-100MGEA 1030 DKK
BLDPBD25810-100MG
4,5,6,7-Tetrahydro-1H-pyrazolo[3,4-c]pyridine dihydrochloride 95%
4,5,6,7-Tetrahydro-1H-pyrazolo[3,4-c]pyridine dihydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₁₁Cl₂N₃
Molekylvægt: 196,08 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07698574
CAS-nummer: 871726-74-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR