4-Iodo-3-(trifluoromethyl)benzoic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD258023-1GEA 187 DKK
BLDPBD258023-1G BLDPBD258023-5G BLDPBD258023-250MG
4-Iodo-3-(trifluoromethyl)benzoic acid 97%
4-Iodo-3-(trifluoromethyl)benzoic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₈H₄F₃IO₂
Molekylvægt: 316,016 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09258947
CAS-nummer: 914636-20-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR