4-Fluoro-3-(trifluoromethoxy)benzoic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD258019-250MGEA 187 DKK
BLDPBD258019-250MG BLDPBD258019-5G BLDPBD258019-1G
4-Fluoro-3-(trifluoromethoxy)benzoic acid 98%
4-Fluoro-3-(trifluoromethoxy)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₄F₄O₃
Molekylvægt: 224,11 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06660161
CAS-nummer: 886496-49-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR