3-Cloro-5-fluorbenzamid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD257855-250MGEA 125 DKK
BLDPBD257855-250MG BLDPBD257855-5G BLDPBD257855-1G
3-Cloro-5-fluorbenzamid 95%
3-Cloro-5-fluorbenzamid

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₅ClFNO
Molekylvægt: 173,57 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03407964
CAS-nummer: 874781-06-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR